Learn More
Darwin Chambers Company Single Door Environmental Chamber 21 cu. ft.
SDP
Catalog No. 501893921
Encompass_Preferred
The KB030-SS is a 23.9 cubic feet single door environmental chamber. Temperature range is 2 to 50 degrees C. Humidity range ambient to 90 percent. This unit comes with 3 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Stainless steel interior. The KB series chambers feature hot gas by-pass technology which controls the temperature of the epoxy coated evaporator for greater uniformity. The ultrasonic humidification system provides excellent humidity control and avoids hot spots seen during chamber mapping of steam boiler equipped chambers.
Catalog No. 50-189-3921 Supplier Darwin Chambers Company Supplier No. KB030SS
Add to Cart
Add to Cart
Request a Quote
The KB030-SS is a 23.9 cubic feet single door environmental chamber. Temperature range is 2 to 50 degrees C. Humidity range ambient to 90 percent. This unit comes with 3 epoxy coated shelves. Exterior is stainless Steel Front continuous coil coated sides with keyed lock. Stainless steel interior. The KB series chambers feature hot gas by-pass technology which controls the temperature of the epoxy coated evaporator for greater uniformity. The ultrasonic humidification system provides excellent humidity control and avoids hot spots seen during chamber mapping of steam boiler equipped chambers.