Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Environmental Chemistry: Nitrates, Phosphates, and Eutrophication
GREENER_CHOICESCI_ED

Catalog No. S96399 Shop All Aldon Products
Change view
Click to view available options
Age:
13 to 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Age
S96399 13 to 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S96399 Supplier Innovating Science™ Supplier No. IS9701
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

  • 50mL nitrate (10% sodium nitrate) solution
  • 50mL phosphate (10% sodium phosphate) solution
  • 50mL nitrate/phosphate (5% sodium nitrate/5% sodium phosphate) solution
  • 50mL control (deionized) water
  • Plastic cups (60)
  • Microscope slides (1 box)
  • Coverslips (1 box)
  • Teacher's guide
  • Student copy-masters

Requires

  • Natural water sample (collected locally)
  • Aged tap water
  • Marking pens

Understand the role of nitrates and phosphates in the process of eutrophication.

After completing this lab, students should understand the difference between point source and nonpoint source pollution and understand the role of nitrates and phosphates in the process of eutrophication.

Observe and examine the effects of nitrates, the effects of phosphates and the effects of a combination of nitrates and phosphates in miniature “ponds.”

Order Info

Class size: 15 groups.

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) To understand the role of nitrates and phosphates in the process of eutrophication
Includes 50mL Nitrate (10% Sodium Nitrate) solution, 50mL Phosphate (10% Sodium Phosphate) solution, 50mL Nitrate/Phosphate (5% Sodium Nitrate/5% Sodium Phosphate) solution, 50mL control (deionized) water, 60 Plastic cups, 1 box microscope slides, 1 box coverslips, teacher's guide student copy masters
Quantity 1
Class Size 15 Lab Groups
Product Type Nitrates, Phosphates and Eutrophication Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.