Learn More
Perkin Elmer US LLC Environmental Hydrocarbons Analysis Pack
Catalog No. 502117250
Encompass_Preferred
Th analysis pack contains: Two 10 mm quartz cuvettes A cuvette holder Environmental Hydrocarbons Touch Apps Software Kit The Touch Apps Software Kit includes the software and SOPs to support routine measurement of oil and grease in water and soil samples by FT-IR spectroscopy. Spectrum Touch software provides a simplified, workflow-driven operating environment; for faster, more reproducible analyses.
Catalog No. 50-211-7250 Supplier Perkin Elmer US LLC Supplier No. L1608006
Th analysis pack contains: Two 10 mm quartz cuvettes A cuvette holder Environmental Hydrocarbons Touch Apps Software Kit The Touch Apps Software Kit includes the software and SOPs to support routine measurement of oil and grease in water and soil samples by FT-IR spectroscopy. Spectrum Touch software provides a simplified, workflow-driven operating environment; for faster, more reproducible analyses.