Learn More
Perkin Elmer US LLC Environmental Hydrocarbons Cyclohexane Application Pack
Catalog No. 502117234
Encompass_Preferred
The Application Pack contains: Two-port Leur Lock %T cell with CaF2 windows Package of 3 spare 0.5 mm Teflon spacers for the %T cell Two 2 mL Leur Lock syringes Liquid cell Leur connected waste tubing Environmental Hydrocarbons Touch Apps Software Kit The Touch Apps Software Kit includes the software and SOPs to support routine measurement of oil and grease in water and soil samples by FT-IR spectroscopy. Spectrum Touch software provides a simplified, workflow-driven operating environment; for faster, more reproducible analyses.
Catalog No. 50-211-7234 Supplier Perkin Elmer US LLC Supplier No. L1608007
Add to Cart
Add to Cart
Request a Quote
The Application Pack contains: Two-port Leur Lock %T cell with CaF2 windows Package of 3 spare 0.5 mm Teflon spacers for the %T cell Two 2 mL Leur Lock syringes Liquid cell Leur connected waste tubing Environmental Hydrocarbons Touch Apps Software Kit The Touch Apps Software Kit includes the software and SOPs to support routine measurement of oil and grease in water and soil samples by FT-IR spectroscopy. Spectrum Touch software provides a simplified, workflow-driven operating environment; for faster, more reproducible analyses.