Learn More
Perkin Elmer US LLC Environmental Internal Standard Mix
Catalog No. 502103708
Encompass_Preferred
Concentration of Element includes: Matrix Content Part Number 5% HNO3/trace HCL 50 µg/mL: Sc N9307808 20 µg/mL: Ge 10 µg/mL: In, lr, Li6, Rh, Tb, Y
Catalog No. 50-210-3708 Supplier Perkin Elmer US LLC Supplier No. N9307808
Concentration of Element includes: Matrix Content Part Number 5% HNO3/trace HCL 50 µg/mL: Sc N9307808 20 µg/mL: Ge 10 µg/mL: In, lr, Li6, Rh, Tb, Y