Learn More
Perkin Elmer US LLC Environmental Standard Kit for NexiON Standard Mode
Catalog No. 502113657
Encompass_Preferred
Concentration of Element includes: Matrix Content Part Number 5% HNO3 1,000 µg/mL: Ca, K, Mg, Na N9307805 5% HNO3 1,000 µg/mL: Al, Fe N9307806 5% HNO3/trace Tartaric Acid/trace HF 100 µg/mL: Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sn, Sr, Ti, Tl, V, Zn N9301721 2% HNO3 100 µg/mL: B, Th, U N9307807 5% HNO3 10 µg/mL: Hg N9300253 5% HNO3 50 µg/mL: Sc, Ge N9308591 10 µg/mL: Rh, In, Tb
Catalog No. 50-211-3657 Supplier Perkin Elmer US LLC Supplier No. N9307118
Concentration of Element includes: Matrix Content Part Number 5% HNO3 1,000 µg/mL: Ca, K, Mg, Na N9307805 5% HNO3 1,000 µg/mL: Al, Fe N9307806 5% HNO3/trace Tartaric Acid/trace HF 100 µg/mL: Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, Pb, Sb, Se, Sn, Sr, Ti, Tl, V, Zn N9301721 2% HNO3 100 µg/mL: B, Th, U N9307807 5% HNO3 10 µg/mL: Hg N9300253 5% HNO3 50 µg/mL: Sc, Ge N9308591 10 µg/mL: Rh, In, Tb