Description
- Available in 10 mL fill volumes
- With high-quality foam swab
- Swab can be securely attached to the cap and makes a convenient 'handle' for great control when sampling
Specifications
Specifications
| Media Type | Letheen Broth |
| Product Type | Prepared Microbiology Tubes |
| Sterility | Non-sterile |
| Includes | Separate Foam Swab |