Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Environmental Toxicity Response in C. elegans Experiment Kit
SDPSCI_ED

Catalog No. s29026 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
9 to Undergraduate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S29026 9 to Undergraduate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S29026 Supplier Edvotek™ Supplier No. 856
Only null left
Add to Cart
Add to Cart

Includes

instructions, C. elegans-normal, C. elegans-Toxicity mutant, petri dishes, NGM medium salts, ReadyPour NGM Agar, E. coli 0P50 Bactobeads™, cell counting chambers, buffer, pipets, sterile loops, tubes and heavy metal compounds.

In this experiment, students will observe and compare the effects of heavy metals found in the environment on normal and mutant strains of Caenorhabditis elegans (C. elegans). Edvotek™ Environmental Toxicity Response in C. elegans Experiment Kit contains five LIVE materials.

  • For 10 Lab Groups
  • Growing Bacteria: overnight; Plating Worms: 15 min.; Worm Growth: 3-4 Days; Toxicity Experiment: 45-60 min.

Specifications

Grade 9 to Undergraduate
Includes Instructions, C. elegans-normal, C. elegans-Toxicity mutant, petri dishes, NGM medium salts, ReadyPour NGM Agar, E. coli 0P50 Bactobeads™, cell counting chambers, buffer, pipets, sterile loops, tubes and heavy metal compounds
Requires Ethanol, timers, microscopes, water bath, covered box, incubator (optional)
Time Range Growing Bacteria: overnight; Plating Worms: 15 minutes; Worm Growth:- 3-4 days; Toxicity Experiment: 45-60 minutes
Content And Storage Some components require refrigerator storage.
Class Size 10 Lab Groups
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.