Learn More
MSC American Formula Wax Stripper, Environmentally Friendly, Low Odor, 5 Gallon Pail
Catalog No. 502837080
Encompass
This product is a highly concentrated, low odor, and environmentally friendly wax stripper. It is designed to quickly penetrate and remove multiple coats of built-up, aged floor finish, including conventional metal interlocked finishes, and works effectively with cold water.
  • Highly concentrated formula
  • Low odor for user comfort
  • Environmentally friendly formulation
  • Effectively removes various floor finishes
  • Works efficiently with cold water
  • Dye and fragrance-free
  • Has a pH range of 11-12
  • Dilution ratio from 5 oz./gallon to 30 oz./gallon
  • Packaged as a 5 gallon pail
Catalog No. 50-283-7080 Supplier MSC Supplier No. 42881672
This product is a highly concentrated, low odor, and environmentally friendly wax stripper. It is designed to quickly penetrate and remove multiple coats of built-up, aged floor finish, including conventional metal interlocked finishes, and works effectively with cold water.
  • Highly concentrated formula
  • Low odor for user comfort
  • Environmentally friendly formulation
  • Effectively removes various floor finishes
  • Works efficiently with cold water
  • Dye and fragrance-free
  • Has a pH range of 11-12
  • Dilution ratio from 5 oz./gallon to 30 oz./gallon
  • Packaged as a 5 gallon pail