Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sonoco™ ThermoSafe Envoy Direct™ Parcel Solutions: 2° to 8°C Coverage

Catalog No. 03600184 Shop All Sonoco ThermoSafe Products
Change view
Click to view available options
Payload:
3.7 L
5.6 L
Dimensions (LxWxH):
10.38 x 8.88 x 3.38 in.
8.88 x 8.88 x 2.88 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Payload Dimensions (LxWxH)
03-600-184 5.6 L 10.38 x 8.88 x 3.38 in.
03-600-182 3.7 L 8.88 x 8.88 x 2.88 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-600-184 Supplier Sonoco™ Supplier No. ED5R1
Add to Cart
Add to Cart

Helps providing cost effective temperature control for regional and national distribution of pharmaceuticals direct from pharmacy. Sonoco™ ThermoSafe Envoy Direct™ Series Parcel Solutions (2° to 8°C Coverage) designed to withstand real-world ambient temperature environments.

  • 2° to 8°C measured throughout the entire product space
  • Ideal for next day delivery
  • Simple and fast assembly drives operational efficiencies
  • Uses minimal components with seasonal conditioning requirements to simplify inventory management
  • Optimized dimensional weight reduces transportation costs
TRUSTED_SUSTAINABILITY

Specifications

Quantity 27 Shippers/Cs.
Length (English) Exterior 10.38 in.
Width (English) Exterior 8.88 in.
Includes Foam shipper with outercarton, corrugate payload box and foam buffer pads
Height (English) Exterior 3.38 in.
Type Parcel Solution
Temperature Range (Metric) 2° to 8°C
Payload 5.6 L
Dimensions (LxWxH) 10.38 x 8.88 x 3.38 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.