Learn More
Medchemexpress LLC HY-70002 500mg Medchemexpress, Enzalutamide CAS:915087-33-1 Purity:>98%
SDP
Catalog No. 502029361
Encompass_Preferred
Medchemexpress, HY-70002 500mg Enzalutamide CAS:915087-33-1 Enzalutamide (MDV3100) is an androgen receptor (AR) antagonist with an IC50 of 36 nM in LNCaP prostate cells. Enzalutamide is an autophagy activator[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9361 Supplier Medchemexpress LLC Supplier No. HY70002500MG
Medchemexpress, HY-70002 500mg Enzalutamide CAS:915087-33-1 Enzalutamide (MDV3100) is an androgen receptor (AR) antagonist with an IC50 of 36 nM in LNCaP prostate cells. Enzalutamide is an autophagy activator[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.