Learn More
Cayman Chemical Enzalutamide, Purity: ≥98%, CAS: 915087-33-1, 50 mg
SDP
Catalog No. 502954684
Encompass_Preferred
A non-steroidal androgen receptor antagonist (IC50 = 36 nM); reduces the efficiency of nuclear translocation of the androgen receptor and impairs both its binding to DNA and the recruitment of coactivators
Catalog No. 50-295-4684 Supplier Cayman Chemical Supplier No. 1159650 MG
A non-steroidal androgen receptor antagonist (IC50 = 36 nM); reduces the efficiency of nuclear translocation of the androgen receptor and impairs both its binding to DNA and the recruitment of coactivators