Learn More
Medchemexpress LLC Enzalutamide | 915087-33-1 | 99.9% | C21H16F4N4O2S | 5 MG
SDP
Catalog No. 5000434701
Encompass_Preferred
Enzalutamide is an analytical standard used in research and analytical applications. It is identified as an androgen receptor (AR) antagonist with an IC50 of 36 nM in LNCaP prostate cells and acts as an autophagy activator.
  • Intended for research and analytical applications
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Used as a reference standard in assays
  • Androgen receptor (AR) antagonist
  • Autophagy activator
  • Commonly used in techniques such as HPLC, GC, and MS
Catalog No. 50-004-34701 Supplier Medchemexpress LLC Supplier No. HY70002R5MG
Enzalutamide is an analytical standard used in research and analytical applications. It is identified as an androgen receptor (AR) antagonist with an IC50 of 36 nM in LNCaP prostate cells and acts as an autophagy activator.
  • Intended for research and analytical applications
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Used as a reference standard in assays
  • Androgen receptor (AR) antagonist
  • Autophagy activator
  • Commonly used in techniques such as HPLC, GC, and MS