Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Enzalutamide | 915087-33-1 | MFCD14155804 | 464.44 g/mol | C21H16F4N4O2S | 10 MG
SDP

Catalog No. 5000298477
Encompass_Preferred

Enzalutamide (Standard) is the analytical reference standard of the androgen receptor antagonist enzalutamide, supplied as a solid powder for research and analytical applications. It is intended for laboratory research use only and includes solubility and storage guidance for preparing stock solutions and in vivo formulations.

  • Analytical standard of enzalutamide for research use only.
  • CAS number 915087-33-1.
  • Chemical formula C21H16F4N4O2S.
  • Molecular weight 464.44 g/mol.
  • Form: powder; available in 10 mg packs.
  • Solubility: ≥50 mg/mL in DMSO; in vivo formulation options and preparation notes provided.
  • Storage: powder -20°C (3 years) or 4°C (2 years); follow recommended temperatures for solutions.

Catalog No. 50-002-98477 Supplier Medchemexpress LLC Supplier No. HY70002R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Enzalutamide (Standard) is the analytical reference standard of the androgen receptor antagonist enzalutamide, supplied as a solid powder for research and analytical applications. It is intended for laboratory research use only and includes solubility and storage guidance for preparing stock solutions and in vivo formulations.

  • Analytical standard of enzalutamide for research use only.
  • CAS number 915087-33-1.
  • Chemical formula C21H16F4N4O2S.
  • Molecular weight 464.44 g/mol.
  • Form: powder; available in 10 mg packs.
  • Solubility: ≥50 mg/mL in DMSO; in vivo formulation options and preparation notes provided.
  • Storage: powder -20°C (3 years) or 4°C (2 years); follow recommended temperatures for solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.