Learn More
Medchemexpress LLC Enzalutamide-based VHL-recruiting androgen receptor degrader (ARCC-4) | 1973403-00-7 | MFCD32858266 | 99.9% | C53H56F3N7O7S2 | 10MG
SDP
Catalog No. 5000261253
Encompass_Preferred
A potent proteolysis-targeting chimera (PROTAC) small molecule that induces targeted proteasomal degradation of the androgen receptor (AR). It demonstrates low-nanomolar degradation activity in cellular assays and is used to probe AR signaling and prostate cancer biology. Molecular formula C53H56F3N7O7S2; molecular weight 1024.18 Da.
  • Low-nanomolar potency for androgen receptor degradation.
  • Mechanism recruits VHL E3 ligase to promote proteasomal degradation.
  • Effective against wild-type and clinically relevant AR mutants in cell models.
  • High purity suitable for in vitro research applications.
  • Available in multiple small-scale pack sizes and as a DMSO solution for convenience.
Catalog No. 50-002-61253 Supplier Medchemexpress LLC Supplier No. HY13049210MG
A potent proteolysis-targeting chimera (PROTAC) small molecule that induces targeted proteasomal degradation of the androgen receptor (AR). It demonstrates low-nanomolar degradation activity in cellular assays and is used to probe AR signaling and prostate cancer biology. Molecular formula C53H56F3N7O7S2; molecular weight 1024.18 Da.
  • Low-nanomolar potency for androgen receptor degradation.
  • Mechanism recruits VHL E3 ligase to promote proteasomal degradation.
  • Effective against wild-type and clinically relevant AR mutants in cell models.
  • High purity suitable for in vitro research applications.
  • Available in multiple small-scale pack sizes and as a DMSO solution for convenience.