Learn More
TARGETMOL CHEMICALS INC Enzalutamide carboxylic acid
SDP
Catalog No. 502876808
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Enzalutamide carboxylic acid is an antagonist of androgen receptor (AR) . Purity 99.17%
Catalog No. 50-287-6808 Supplier TARGETMOL CHEMICALS INC Supplier No. T82175MG
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Enzalutamide carboxylic acid is an antagonist of androgen receptor (AR) . Purity 99.17%