Learn More
Medchemexpress LLC Enzalutamide-d6 | 1443331-94-9 | 99.9% | 470.47 g/mol | C21H10D6F4N4O2S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY70002S110MG
Encompass_Preferred
Enzalutamide-d6 is a deuterium-labeled analogue of enzalutamide supplied for analytical and research use as a tracer or internal standard in quantitative workflows. The material is provided as a white to off-white solid with a reported molecular weight of 470.47 g/mol and high purity (~99.9%).
  • Deuterium-labeled internal standard (d6) for quantitative analysis (NMR, GC-MS, LC-MS).
  • Suitable as a tracer for analytical method development and validation.
  • Reported androgen receptor antagonist reference activity (IC50 36 nM in LNCaP cells).
  • High purity and consistent lot reporting support analytical accuracy.
  • Recommended storage: sealed at 4°C; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Available in small pack sizes for analytical use, including a 10 mg pack.
Catalog No. 50-002-98646