Learn More
Medchemexpress LLC Enzalutamide-d6 (MDV3100-d6) | 1443331-94-9 | 99.9% | C21H10D6F4N4O2S | 5 MG
SDP
Catalog No. 5000433580
Encompass_Preferred
Enzalutamide-d6 (MDV3100-d6) is a deuterium labeled Enzalutamide (MDV3100), acting as an androgen receptor (AR) antagonist with an IC50 of 36 nM in LNCaP prostate cells.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Contains stable heavy isotopes incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-33580 Supplier Medchemexpress LLC Supplier No. HY70002S15MG
Enzalutamide-d6 (MDV3100-d6) is a deuterium labeled Enzalutamide (MDV3100), acting as an androgen receptor (AR) antagonist with an IC50 of 36 nM in LNCaP prostate cells.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Contains stable heavy isotopes incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.