Learn More
Medchemexpress LLC ENZALUTAMIDE (MDV3100); A100MG
SDP
Supplier:  Medchemexpress LLC HY70002100MG
Encompass_Preferred
Enzalutamide (MDV3100); androgen-receptor (AR) antagonist; 100 mg
Catalog No. NC1448643