Learn More
Selleck Chemical LLC Enzastaurin S1055-10mM/1mL
SDP
Catalog No. 503108522
Encompass_Preferred
Enzastaurin is a potent PKC selective inhibitor with IC50 of 6 nM in cell-free assays 6- to 20-fold selectivity against PKC PKC and PKC Phase 3
Catalog No. 50-310-8522 Supplier Selleck Chemical LLC Supplier No. S105510MM/1ML
Enzastaurin is a potent PKC selective inhibitor with IC50 of 6 nM in cell-free assays 6- to 20-fold selectivity against PKC PKC and PKC Phase 3