Learn More
TARGETMOL CHEMICALS INC ENZASTAURIN 50MG
SDP
Catalog No. 502576544
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Enzastaurin (LY317615) (LY317615) is an effective PKC(beta) selective inhibitor (IC50 6 nM) 6- to 20-fold selectivity against PKC(alpha)/(gamma)/(epsilon). purity: 98%
Catalog No. 50-257-6544 Supplier TARGETMOL CHEMICALS INC Supplier No. T628050MG
Also available in 5 mg 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Enzastaurin (LY317615) (LY317615) is an effective PKC(beta) selective inhibitor (IC50 6 nM) 6- to 20-fold selectivity against PKC(alpha)/(gamma)/(epsilon). purity: 98%