Learn More
Medchemexpress LLC HY-10342 5mg , Enzastaurin CAS:170364-57-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY103425MG
Encompass_Preferred
HY-10342 5mg Enzastaurin CAS: 170364-57-5 Enzastaurin (LY317615) is a potent and selective PKCβ inhibitor with an IC50 of 6 nM, showing 6- to 20-fold selectivity over PKCα, PKCγ and PKCε. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9437