Learn More
Apexbio Technology LLC Enzastaurin (LY317615) 170364-57-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063042
Encompass_Preferred
Enzastaurin (LY317615 CAS 170364-57-5) is an orally bioavailable ATP-competitive inhibitor selective for protein kinase C beta (PKC ) a member of the serine-threonine kinase family implicated in tumor growth and progression By inhibiting PKC (IC50 6 nM) enzastaurin decreases phosphorylation and signaling of downstream effectors including ribosomal protein S6 GSK3 and AKT In cellular assays enzastaurin reduces proliferation and induces apoptosis across diverse tumor types (e g colon cancer HCT-116 glioblastoma U87MG) In vivo studies demonstrate antitumor efficacy in xenograft models highlighting its utility for oncology research
Catalog No. 50-000-63042 Supplier Apexbio Technology LLC Supplier No. A16705.1
Enzastaurin (LY317615 CAS 170364-57-5) is an orally bioavailable ATP-competitive inhibitor selective for protein kinase C beta (PKC ) a member of the serine-threonine kinase family implicated in tumor growth and progression By inhibiting PKC (IC50 6 nM) enzastaurin decreases phosphorylation and signaling of downstream effectors including ribosomal protein S6 GSK3 and AKT In cellular assays enzastaurin reduces proliferation and induces apoptosis across diverse tumor types (e g colon cancer HCT-116 glioblastoma U87MG) In vivo studies demonstrate antitumor efficacy in xenograft models highlighting its utility for oncology research