Learn More
Apexbio Technology LLC Enzastaurin (LY317615), 5mg. Cas: 170364-57-5 MFCD: MFCD11040980. PKC beta inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC A16705
Encompass_Preferred
Enzastaurin is an ATP-competitive, selective oral inhibitor of protein kinase C with IC50 value of 6 nM [1].Protein kinase C (PKC) is a family of serine-threonine protein kinases that have been proved to play critical roles in the formation and progression of tumor cells. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4703