Learn More
NANOPROBES INC ENZMET IHC/ISH HRP KIT
Catalog No. NC1452880
Encompass
150 slides,30mL EnzMet™ metallographic substrate for horseradish peroxidase. Formulated for imunohistochemistry (IHC) and in situ hybridization (ISH) applications.
Catalog No. NC1452880 Supplier NANOPROBES INC Supplier No. 600130ML
150 slides,30mL EnzMet™ metallographic substrate for horseradish peroxidase. Formulated for imunohistochemistry (IHC) and in situ hybridization (ISH) applications.