Learn More
Siemens Healthineers Enzygnost™ F1+2 (Monoclonal)
Catalog No. 23044668 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
192 Tests/Kit
Catalog No. Quantity
23-044-668 192 Tests/Kit
1 options
This item is not returnable. View return policy
Catalog No. 23-044-668 Supplier Siemens Healthineers Supplier No. 10445978
Add to Cart
Add to Cart
This item is not returnable. View return policy
Quantitative determination of prothrombin fragment F 1+2 in human plasma.
Enzygnost™ F1+2 (monoclonal) is an enzyme immunoassay for the quantitative determination of the human prothrombin fragment F1+2 in plasma. Measurement of F1+2 is used as an aid in the diagnosis, monitoring, and evaluating of acquired or hereditary blood coagulation disorders. It is indicated as an aid in assessing risk of thrombosis and in monitoring efficacy of anticoagulant therapy.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Kit
For Use With (Application) Prothrombin Time (PT) Testing
Quantity 192 Tests/Kit
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov