Learn More
Siemens Healthineers Enzygnost™ TAT Micro Kit
Catalog No. 23044704 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
192 Tests/Kit
Catalog No. Quantity
23-044-704 192 Tests/Kit
1 options
This item is not returnable. View return policy
Catalog No. 23-044-704 Supplier Siemens Healthineers Supplier No. 10446632
Add to Cart
Add to Cart
This item is not returnable. View return policy
Quantitative determination of thrombin/antithrombin III complex (TAT) in human plasma.
Enzygnost™ TAT micro is an enzyme immunoassay for the determination of human thrombin/antithrombin III complex in plasma as an aid in the diagnosis and monitoring of thrombosis and related conditions.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Kit
For Use With (Application) Thrombin/Antithrombin III Complex (TAT) Testing
Quantity 192 Tests/Kit
WARNING: Cancer - www.P65Warnings.ca.gov