Learn More
QA Bio Inc Enzymatic CarboRelease Kit
Supplier:  QA Bio Inc KEDG01
Encompass
Includes the enzymes and buffers required to deglycosylate glycoproteins, removing all N-linked oligosaccharides and many O-linked sugars. • The enzymes are packaged separately in individual 20 microlitre vials. This gives researchers the flexibility to characterize the glycans attached to their glycoprotein more fully than with our mixture of enzymes, KE-DGMX. • Appropriate use of individual glycosidases allow the determination of sialylation (use of Neuraminidase only) and the specific presence of N-linked glycans (using PNGase F only) and O-linked glycans (using Neuraminidase, Beta-Galactosidase, O-Glycosidase, and β-N-acetylglucosaminidase). • Includes 20 microlitres each of the following enzymes: PNGase F - O-Glycosidase - Neuraminidase - Beta-Galactosidase - β-N-acetylglucosaminidase • Other Supplied Reagents: Reaction buffer - Denaturation Solution - Triton X -Bovine Fetuin (control)
Catalog No. 50-223-6690