Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enzymatic Digest Casein Hydrolysate

Catalog No. AAJ12855Q1 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
11.3 kg (25 lb.)
2.2 kg (5 lb.)
453.5 g (1 lb.)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ12855Q1 11.3 kg (25 lb.)
AAJ12855P2 453.5 g (1 lb.)
AAJ12855P5 2.2 kg (5 lb.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ12855Q1 Supplier Thermo Scientific Supplier No. J12855Q1
Add to Cart

missing translation for 'validMainframeMsg'

Enzymatic digest of casein.

  • Synonym: Peptones
  • pH: 6.6 to 7.5
  • CAS: 65072-00-6
  • MDLNumber: MFCD00282824
TRUSTED_SUSTAINABILITY

Specifications

Name Casein hydrolysate
pH Range 6.6 to 7.5
Quantity 11.3 kg (25 lb.)
Packaging Plastic bottle
Form Powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.