Learn More
NEOGEN Enzymatic Digest of Soy No. 2, 500 g
SDP
Catalog No. 502015795
Encompass_Preferred
Enzymatic Digest of Soy No. 2 is an enzymatic digest of soybean meal for use in preparing microbiological culture media in a laboratory setting. Enzymatic Digest of Soy No. 2 is not intended for use in the diagnosis of disease or other conditions in humans.Enzymatic Digest of Soy No. 2 is an enzymatic hydrolysate of soybean meal prepared under controlled conditions for use in microbiological procedures. Enzymatic Digest of Soy No. 2 is recommended for use in media for the cultivation of a large variety of organisms, including fungi and microbiological assay media. The nitrogen source in Enzymatic Digest of Soy No. 2 contains naturally occurring, high concentrations of vitamins and carbohydrates of soybean. Due to its high carbohydrate content, Enzymatic Digest of Soy No. 2 cannot be used for fermentation studies.For more information visit https://www.neogen.com/solutions/Microbiology/enzymatic-digest-of-soy-number-2
Catalog No. 50-201-5795 Supplier NEOGEN Supplier No. 700003564
Add to Cart
Add to Cart
Enzymatic Digest of Soy No. 2 is an enzymatic digest of soybean meal for use in preparing microbiological culture media in a laboratory setting. Enzymatic Digest of Soy No. 2 is not intended for use in the diagnosis of disease or other conditions in humans.Enzymatic Digest of Soy No. 2 is an enzymatic hydrolysate of soybean meal prepared under controlled conditions for use in microbiological procedures. Enzymatic Digest of Soy No. 2 is recommended for use in media for the cultivation of a large variety of organisms, including fungi and microbiological assay media. The nitrogen source in Enzymatic Digest of Soy No. 2 contains naturally occurring, high concentrations of vitamins and carbohydrates of soybean. Due to its high carbohydrate content, Enzymatic Digest of Soy No. 2 cannot be used for fermentation studies.For more information visit https://www.neogen.com/solutions/Microbiology/enzymatic-digest-of-soy-number-2