Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diazyme Laboratories Enzymatic Homocysteine 5 Level Calibrator

Catalog No. 23700538
Encompass
Encompass_Preferred
Change view
Click to view available options
Type:
Diagnostic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-700-538 Diagnostic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-700-538 Supplier Diazyme Laboratories Supplier No. DZ568ACA5
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Diazyme 5 Level Homocysteine Calibrator Set is for the testing of Homocysteine in serum.

  • A liquid stable system, which provides a fast, accurate cost effective alternative to cumbersome and expensive immunochemical methods
  • A choice of convenient instrument specific packaging options eliminate reagent transfer for Roche™ Hitachi, Siemens Dimension™, and Beckman Synchron CX, LX and DXC analyzers
  • Accurate and reliable, the assay features excellent correlation with HPLC and Immunochemical methods with a linear range of 3-50 μmole/L and intra and inter %CV values of < 5.9%
  • The assay has no significant interference from lipemic and hemolytic substances and eliminates the carry-over interference with iron or lipase reagents that can occur with other methods
  • More significantly, the assay is not interfered by the endogenous cystathionine and therefore can be used with confidence in patients with renal disease

Specifications

Certifications/Compliance 510(k) Cleared, CE, Health Canada Registered
Content And Storage 2°C to 8°C. DO NOT FREEZE. Note: Store CALIBRATOR tightly capped when not in use
Description ENZYMATIC HOMOCYSTEIN 5 PT CALIBRATION SET
CE Marker Yes
Detectable Analytes Homocystein
Format 5 x 3 mL Vial
Type Diagnostic
Includes 5 vials
Stability Closed Vial: Stability is up to the expiration date on label when stored at 2 to 8°C. Open Vial: Stability is up to 60 days when stored at 2 to 8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.