Learn More
MOLECULAR DEPOT LLC Enzyme Acceptor Buffer - 50 mL
Catalog No. 5000013430
Encompass_Preferred
Beta-galactosidase Enzyme Acceptor Stabilization Buffer (50 mL) consists of a buffer specifically optimized to stabilize the beta-galactosidase fragment known as enzyme acceptor (Omega fragment) The buffer can extend the shelf life of the enzyme acceptor to up to 12 months at 2-8C Without this buffer the enzyme acceptor is highly instable and loses activity at a high rate For in vitro research use only
Catalog No. 50-000-13430 Supplier MOLECULAR DEPOT LLC Supplier No. B2010002
Beta-galactosidase Enzyme Acceptor Stabilization Buffer (50 mL) consists of a buffer specifically optimized to stabilize the beta-galactosidase fragment known as enzyme acceptor (Omega fragment) The buffer can extend the shelf life of the enzyme acceptor to up to 12 months at 2-8C Without this buffer the enzyme acceptor is highly instable and loses activity at a high rate For in vitro research use only