Learn More
Eurofins Discoverx CK1 delta Kinase Enzyme Activity Assay Kit
Catalog No. 5000455573
Encompass
The CK1delta Kinase Enzyme Activity Assay Kit provides a fully validated, ready-to-use biochemical assay built on the trusted ADP Hunter™ Plus platform from DiscoverX, enabling accurate assessment of kinase inhibitor potency. This comprehensive kit includes recombinant protein, substrate, ATP, detection reagents, and assay plates for precise measurement of ADP formation.
Catalog No. 50-004-55573 Supplier Eurofins Discoverx Supplier No. 44520L
The CK1delta Kinase Enzyme Activity Assay Kit provides a fully validated, ready-to-use biochemical assay built on the trusted ADP Hunter™ Plus platform from DiscoverX, enabling accurate assessment of kinase inhibitor potency. This comprehensive kit includes recombinant protein, substrate, ATP, detection reagents, and assay plates for precise measurement of ADP formation.