Learn More
Eurofins Discoverx LRRK2 Kinase Enzyme Activity Assay Kit
Catalog No. 5000455699
Encompass
The LRRK2 Kinase Enzyme Activity Assay Kit provides a fully validated, ready-to-use biochemical assay built on the trusted ADP Hunter™ Plus platform from DiscoverX, enabling accurate assessment of kinase inhibitor potency. This comprehensive kit includes recombinant protein, substrate, ATP, detection reagents, and assay plates for precise measurement of ADP formation.
Catalog No. 50-004-55699 Supplier Eurofins Discoverx Supplier No. 44919L
The LRRK2 Kinase Enzyme Activity Assay Kit provides a fully validated, ready-to-use biochemical assay built on the trusted ADP Hunter™ Plus platform from DiscoverX, enabling accurate assessment of kinase inhibitor potency. This comprehensive kit includes recombinant protein, substrate, ATP, detection reagents, and assay plates for precise measurement of ADP formation.