Learn More
Quanterix Inc ENZYME AND SUBSTRATE KIT CONTA
SDP
Supplier:  Quanterix Inc 101361
Encompass
The Simoa Homebrew Assay Development Kit is comprised of beads, buffers, labels and detailed protocols for preparing custom digital immunoassay reagents for research that are capable of greatly exceeding the sensitivity of conventional immunoassays. The Simoa Homebrew Assay Development Kit has been configured for flexibility, providing materials for multiple reagent batches and step-by-step instructions to improve assay performance with a given antigen/antibody combination. The comprehensive Simoa Homebrew Assay Development Kit includes assay development supplies as well as instructional materials to be downloaded from the Quanterix(TM) website. REAGENTS AND MATERIALS INCLUDED Simoa Homebrew Assay Development Kit (101354) Homebrew Carboxylated Paramagnetic Beads 1 bottle (5 mL) Approximately 1010 microbeads in phosphate buffer. Preservative: Sodium azide. Homebrew Bead Conjugation Buffer 3 bottles (110 mL ea.) 2-(N-morpholino)ethanesulfonate (MES) buffer. Preservative: ProClin 300. Hom
Catalog No. NC0789719