Learn More
Biochain Institute Inc Enzyme Alpha-Amylase Assay Kit, 100 assays/PK
SDP
Catalog No. 50180854
Encompass_Preferred
BioChain's Alpha-amylase Assay Kits include two types: Type I: An insoluble dye-coupled substrate amylose azure is cleaved by alpha-amylase into soluble colored products. The color intensity, measured at 595 nm, is proportionate to the enzyme activity in the sample. Type II: Two steps are involved: (1). Alpha-amylase in the sample hydrolyzes starch and the product is rapidly converted to glucose by alpha-glucosidase and hydrogen peroxide by glucose oxidase; (2). hydrogen peroxide concentration is determined with a colorimetric reagent.
Catalog No. 50-180-854 Supplier Biochain Institute Inc Supplier No. Z5030046
BioChain's Alpha-amylase Assay Kits include two types: Type I: An insoluble dye-coupled substrate amylose azure is cleaved by alpha-amylase into soluble colored products. The color intensity, measured at 595 nm, is proportionate to the enzyme activity in the sample. Type II: Two steps are involved: (1). Alpha-amylase in the sample hydrolyzes starch and the product is rapidly converted to glucose by alpha-glucosidase and hydrogen peroxide by glucose oxidase; (2). hydrogen peroxide concentration is determined with a colorimetric reagent.