Learn More
Meridian Life Science Enzyme Dilution Buffer 10x, 100ML
SDP
Catalog No. 5000856236
Encompass_Preferred
Meridian Bioscience Enzyme Dilution Buffer 10x is a glycerol-free dilution buffer, for the dilution of enzymes to reaction concentration.
Catalog No. 50-008-56236 Supplier Meridian Life Science Supplier No. MDX080100
Meridian Bioscience Enzyme Dilution Buffer 10x is a glycerol-free dilution buffer, for the dilution of enzymes to reaction concentration.