Learn More
Meridian Life Science Enzyme Dilution Buffer 10x, 5ML
SDP
Catalog No. 5000856406
Encompass_Preferred
Meridian Bioscience Enzyme Dilution Buffer 10x is a glycerol-free dilution buffer, for the dilution of enzymes to reaction concentration.
Catalog No. 50-008-56406 Supplier Meridian Life Science Supplier No. MDX0805
Meridian Bioscience Enzyme Dilution Buffer 10x is a glycerol-free dilution buffer, for the dilution of enzymes to reaction concentration.