Learn More
Meridian Life Science Enzyme Dilution Buffer 1x, 100ML
SDP
Catalog No. 5000856291
Encompass_Preferred
Meridian Bioscience Enzyme Dilution Buffer 1x is a glycerol containing dilution buffer, for the dilution of enzymes to reaction concentration.
Catalog No. 50-008-56291 Supplier Meridian Life Science Supplier No. MDX078100
Meridian Bioscience Enzyme Dilution Buffer 1x is a glycerol containing dilution buffer, for the dilution of enzymes to reaction concentration.