Learn More
Meridian Life Science Enzyme Dilution Buffer 1x, 5ML
SDP
Catalog No. 5000856503
Encompass_Preferred
Meridian Bioscience Enzyme Dilution Buffer 1x is a glycerol containing dilution buffer, for the dilution of enzymes to reaction concentration.
Catalog No. 50-008-56503 Supplier Meridian Life Science Supplier No. MDX0785
Meridian Bioscience Enzyme Dilution Buffer 1x is a glycerol containing dilution buffer, for the dilution of enzymes to reaction concentration.