Learn More
MOLECULAR DEPOT LLC Enzyme Donor Buffer - 50 mL
Catalog No. 5000013329
Encompass_Preferred
Beta-galactosidase Enzyme Donor Stabilization Buffer (50 mL) consists of a buffer specifically optimized to stabilize the beta-galactosidase fragment known as enzyme donor (Alpha fragment) The buffer can extend the shelf life of the enzyme donor to up to 12 months at 2-8C Without this buffer the enzyme donor is highly instable and loses activity at a high rate For in vitro research use only
Catalog No. 50-000-13329 Supplier MOLECULAR DEPOT LLC Supplier No. B2010001
Beta-galactosidase Enzyme Donor Stabilization Buffer (50 mL) consists of a buffer specifically optimized to stabilize the beta-galactosidase fragment known as enzyme donor (Alpha fragment) The buffer can extend the shelf life of the enzyme donor to up to 12 months at 2-8C Without this buffer the enzyme donor is highly instable and loses activity at a high rate For in vitro research use only