Learn More
Verichem Laboratories ENZYME ER VERIFIER
Catalog No. NC9014986
Encompass_Preferred
Enzyme ER Verifier Kit. aEc Protein-based, liquid, ready-to-use aEc Intended for calibration verification of serum kinetic assays aEc Known linear relation stated in product insert aEc Stable 14 months from date of manufacture aEc Specifically designed for extended range systems aEc assay values included for Beckman aU Chemistry Systems
Catalog No. NC9014986 Supplier Verichem Laboratories Supplier No. 9410
Enzyme ER Verifier Kit. aEc Protein-based, liquid, ready-to-use aEc Intended for calibration verification of serum kinetic assays aEc Known linear relation stated in product insert aEc Stable 14 months from date of manufacture aEc Specifically designed for extended range systems aEc assay values included for Beckman aU Chemistry Systems