Learn More
STEMCELL Technologies ReLeSR™, Size: 500 mL
SDP
Catalog No. 502069324
Encompass
ReLeSR™ is an enzyme-free reagent for dissociation and passaging of human embryonic stem (ES) or induced pluripotent stem (iPS) cells as aggregates without manual selection or scraping. Passaging human ES/iPS cells with ReLeSR™ easily generates optimally-sized aggregates, while eliminating the hassle and variability associated with manual manipulation. By eliminating the need for scraping, ReLeSR™ enables the use of culture flasks and other closed vessels, thus facilitating culture scale-up and automation. ReLeSR™, 500 mL, is now also manufactured following relevant cGMPs under a certified quality management system to ensure the highest quality and consistency for reproducible results.To request a Letter of Authorization (LOA) for ReLeSR’s (Catalog #100-0484) Drug Master File, click here (https://www.stemcell.com/loa-request-dmf).
Catalog No. 50-206-9324 Supplier STEMCELL Technologies Supplier No. 1000484
ReLeSR™ is an enzyme-free reagent for dissociation and passaging of human embryonic stem (ES) or induced pluripotent stem (iPS) cells as aggregates without manual selection or scraping. Passaging human ES/iPS cells with ReLeSR™ easily generates optimally-sized aggregates, while eliminating the hassle and variability associated with manual manipulation. By eliminating the need for scraping, ReLeSR™ enables the use of culture flasks and other closed vessels, thus facilitating culture scale-up and automation. ReLeSR™, 500 mL, is now also manufactured following relevant cGMPs under a certified quality management system to ensure the highest quality and consistency for reproducible results.To request a Letter of Authorization (LOA) for ReLeSR’s (Catalog #100-0484) Drug Master File, click here (https://www.stemcell.com/loa-request-dmf).