Learn More
TARGETMOL CHEMICALS INC EO-1606 1MG
SDP
Catalog No. 502878064
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. EO-1606 is a p38MAP kinase inhibitor with anti-inflammatory activity and may be used in studies of Alzheimer;s disease and dermatitis. Purity 99.27%
Catalog No. 50-287-8064 Supplier TARGETMOL CHEMICALS INC Supplier No. T316451MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. EO-1606 is a p38MAP kinase inhibitor with anti-inflammatory activity and may be used in studies of Alzheimer;s disease and dermatitis. Purity 99.27%