Learn More
SPS Medical EO GAS IND STRIPS 4IN 250PK
Catalog No. NC9019015
Encompass_Preferred
Indicator strips provide distinct color change when exposed to EO Gas cycles. Eight inch strips can be used for larger packs or torn in half at perforation for smaller packs.
Catalog No. NC9019015 Supplier SPS Medical Supplier No. GIS250
Indicator strips provide distinct color change when exposed to EO Gas cycles. Eight inch strips can be used for larger packs or torn in half at perforation for smaller packs.