Learn More
Hach Company EO Sterilized Petri Dish w/Pad
Supplier:  Hach Company 25248000
Encompass
Ethylene oxide (EO) sterilized Petri dish with pad (47 mm) Sold as a convenient 150 pack and recommended for use with Hach s m-ColiBlue24 and other micro broths Manufactured by Millipore
Catalog No. 50-002-07173