Learn More
ATCC EOC 13.31; Brain Microglia; Mouse (Mus musculus)
Catalog No. 502383611
Encompass
EOC 13.31 is a microglial cell line that was isolated from the brain of a 10-day-old, female mouse. This cell line was deposited by WS Walker and can be used in neuroscience research.
Catalog No. 50-238-3611 Supplier ATCC Supplier No. CRL2468
EOC 13.31 is a microglial cell line that was isolated from the brain of a 10-day-old, female mouse. This cell line was deposited by WS Walker and can be used in neuroscience research.