Learn More
Medchemexpress LLC HY-16025 2mg Medchemexpress, EOC317 CAS:939805-30-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY160252MG
Encompass_Preferred
Medchemexpress, HY-16025 2mg EOC317 CAS:939805-30-8 EOC317 (ACTB-1003) is an oral kinase inhibitor with IC50s of 6, 2 and 4 nM for FGFR1, VEGFR2 and Tie-2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1865