Learn More
ADINSTRUMENTS EOG Kit, horizontal/vertical eye movement tracking, includes EOG Pod, shielded lead wires, 100 disposable ECG electrodes
SDP
Catalog No. NC3705487
Encompass
The ADInstruments PTK15 EOG Kit is engineered for precise recording and analysis of both horizontal and vertical eye movements. It's ideal for scientific investigations into ocular phenomena such as slow-tracking and saccades. This kit is designed for immediate deployment in experiments targeting eye movement dynamics.
  • Measurement capability: Horizontal and vertical eye movement
  • Primary application: Eye position tracking
  • Research applications: Slow-tracking, saccades
  • Kit contents include: EOG Pod, shielded lead wires, disposable ECG electrodes (100)
  • Manufacturer: ADInstruments
Catalog No. NC3705487 Supplier ADINSTRUMENTS Supplier No. PTK15
Add to Cart
Add to Cart
The ADInstruments PTK15 EOG Kit is engineered for precise recording and analysis of both horizontal and vertical eye movements. It's ideal for scientific investigations into ocular phenomena such as slow-tracking and saccades. This kit is designed for immediate deployment in experiments targeting eye movement dynamics.
  • Measurement capability: Horizontal and vertical eye movement
  • Primary application: Eye position tracking
  • Research applications: Slow-tracking, saccades
  • Kit contents include: EOG Pod, shielded lead wires, disposable ECG electrodes (100)
  • Manufacturer: ADInstruments