Learn More
Grainger EOL Resistor PK2
Catalog No. 502767172
Encompass_Preferred
The EOL Resistor 2PK from Grainger is a pack of two end-of-line resistors. These resistors are used in fire alarm systems to provide supervision of the wiring. They feature nickel-plated brass terminals and have a resistance value of 4.7k ohms. The resistors have a power rating of 1 watt. This 2-pack provides an economical way to obtain the resistors needed for installing and maintaining fire alarms. The EOL resistors are UL listed and are 1/4 watt.
Catalog No. 50-276-7172 Supplier Grainger Supplier No. 16X396
The EOL Resistor 2PK from Grainger is a pack of two end-of-line resistors. These resistors are used in fire alarm systems to provide supervision of the wiring. They feature nickel-plated brass terminals and have a resistance value of 4.7k ohms. The resistors have a power rating of 1 watt. This 2-pack provides an economical way to obtain the resistors needed for installing and maintaining fire alarms. The EOL resistors are UL listed and are 1/4 watt.